Kalium(4-tert-butylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium (4-(tert-butyl)phenyl)trifluoroborate

TB-3255-1G TB-3255-5G TB-3255-25G
COBBTB-3255-1GEA 26.2 EUR
COBBTB-3255-1G COBBTB-3255-5G COBBTB-3255-25G
Kalium(4-tert-butylphenyl)trifluoroborat
Kalium(4-tert-butylphenyl)trifluoroborat
Formel: C₁₀H₁₃BF₃K
Molecular Weight: 240,11 g/mol
MDL: MFCD06659940
CAS-Nummer: 423118-47-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR