4-Brom-2-chlor-5-methylanisole

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-5-chloro-4-methoxy-2-methylbenzene

OT-0713-5G OT-0713-1G
COBBOT-0713-5GEA 747 EUR
COBBOT-0713-5G COBBOT-0713-1G
4-Brom-2-chlor-5-methylanisole
4-Brom-2-chlor-5-methylanisole
Formel: C₈H₈BrClO
MDL: MFCD11504949
CAS-Nummer: 179897-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR