3-Brom-5-(methylthio)benzotrifluorid
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-5-trifluoromethylthioanisole
OT-0573-5G
OT-0573-1G
COBBOT-0573-5GEA
796
EUR
COBBOT-0573-5G
COBBOT-0573-1G
3-Brom-5-(methylthio)benzotrifluorid
3-Brom-5-(methylthio)benzotrifluorid
Formel:
C₈H₆BrF₃S |
MDL:
MFCD11504946 CAS-Nummer: 1072944-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...