3-Brom-4-methoxy-5-methylanilin

Lieferant: COMBI-BLOCKS

AN-1568-5G AN-1568-1G
COBBAN-1568-5GEA 748 EUR
COBBAN-1568-5G COBBAN-1568-1G
3-Brom-4-methoxy-5-methylanilin
3-Brom-4-methoxy-5-methylanilin
Formel: C₈H₁₀BrNO
MDL: MFCD16618926
CAS-Nummer: 1215205-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR