2-Chlor-5-methyl-4-nitroanisol

Lieferant: COMBI-BLOCKS

AN-1534-25G AN-1534-1G AN-1534-5G
COBBAN-1534-25GEA 0 EUR
COBBAN-1534-25G COBBAN-1534-1G COBBAN-1534-5G
2-Chlor-5-methyl-4-nitroanisol
2-Chlor-5-methyl-4-nitroanisol
Formel: C₈H₈ClNO₃
MDL: MFCD14525739
CAS-Nummer: 62492-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR