3,5-Dihydroxy-4-methylbenzoesäure ≥97%

Lieferant: Thermo Scientific
Achtung

A19196.03 A19196.06 A19196.14
A19196.03EA 32.2 EUR
A19196.03 A19196.06 A19196.14
3,5-Dihydroxy-4-methylbenzoesäure ≥97%
3,5-Dihydroxy-4-methylbenzoic acid

Purity: 97% CAS No.: 28026-96-2 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 266-207° MDL No.: MFCD00002514

Formel: C₈H₈O₄
Molecular Weight: 168,15 g/mol
Schmelzpunkt: 266…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002514
CAS-Nummer: 28026-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR