2-Isopropoxy-5-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Isopropoxy-5-(trifluoromethyl)phenyl)boronic acid, 2-Isopropoxy-5-(trifluoromethyl)benzeneboronic acid, 2-Isopropoxy-5-(trifluoromethyl)phenylboronic acid
BB-2792-5G
BB-2792-1G
COBBBB-2792-5GEA
884
EUR
COBBBB-2792-5G
COBBBB-2792-1G
2-Isopropoxy-5-(trifluormethyl)phenylboronsäure
2-Isopropoxy-5-(trifluormethyl)phenylboronsäure
Formel:
C₁₀H₁₂BF₃O₃ Molecular Weight: 248,0100 g/mol |
MDL:
MFCD07363786 CAS-Nummer: 850593-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...