2-Ethoxy-5-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 2-Ethoxy-5-trifluoromethylphenylboronic acid
BB-2790-5G
BB-2790-1G
COBBBB-2790-5GEA
451
EUR
COBBBB-2790-5G
COBBBB-2790-1G
2-Ethoxy-5-(trifluormethyl)phenylboronsäure
2-Ethoxy-5-(trifluormethyl)phenylboronsäure
Formel:
C₉H₁₀BF₃O₃ Molecular Weight: 233,98 g/mol |
MDL:
MFCD06801695 CAS-Nummer: 850593-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...