Kalium(2-fluor-4-(trifluormethyl)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium 2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate

TB-3559-1G TB-3559-5G
COBBTB-3559-1GEA 98.3 EUR
COBBTB-3559-1G COBBTB-3559-5G
Kalium(2-fluor-4-(trifluormethyl)phenyl)trifluoroborat
Potassium-2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
Formel: C₇H₃BF₇K
Molecular Weight: 269.9969 g/mol
MDL: MFCD07781236
CAS-Nummer: 1150655-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR