2-(Trifluormethyl)benzoesäurehydrazid

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluoromethyl)benzohydrazide, 2-(Trifluoromethyl)benzhydrazide

008429-1G 008429-5G
FLUO008429-1GEA 78.9 EUR
FLUO008429-1G FLUO008429-5G
2-(Trifluormethyl)benzoesäurehydrazid
2-(Trifluoromethyl)benzoic acid hydrazide
Formel: C₈H₇F₃N₂O
Molecular Weight: 204,150 g/mol
MDL: MFCD00221473
CAS-Nummer: 344-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR