4-(Trifluormethyl)benzoesäurehydrazid

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluoromethyl)benzoylhydrazine, 4-(Trifluoromethyl)benzhydrazide, 4-(Trifluoromethyl)benzoic Hydrazide, 4-(Trifluoromethyl)benzohydrazide

CFF690.5 008431-5G 008431-25G
CHMPCFF690.5EA 268 EUR
CHMPCFF690.5 FLUO008431-5G FLUO008431-25G
4-(Trifluormethyl)benzoesäurehydrazid
4-(Trifluoromethyl)benzoic acid hydrazide
Formel: C₈H₇F₃N₂O
Molecular Weight: 204.15 g/mol
MDL: MFCD00051703
CAS-Nummer: 339-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR