4-(Dimethylamino)-3-nitrobenzotrifluorid

Lieferant: FLUOROCHEM

009151-1G 009151-5G
FLUO009151-1GEA 88.1 EUR
FLUO009151-1G FLUO009151-5G
4-(Dimethylamino)-3-nitrobenzotrifluorid
4-(Dimethylamino)-3-nitrobenzotrifluorid
Formel: C₉H₉F₃N₂O₂
MDL: MFCD00236670
CAS-Nummer: 40700-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR