2-(p-Toluolsulfonyl)ethanol

Lieferant: FLUOROCHEM

Synonyme: 2-Tosylethanol

008058-5G 008058-25G
FLUO008058-5GEA 139 EUR
FLUO008058-5G FLUO008058-25G
2-(p-Toluolsulfonyl)ethanol
2-(p-Toluolsulfonyl)ethanol
Formel: C₉H₁₂O₃S
Molecular Weight: 200,2600 g/mol
MDL: MFCD00041757
CAS-Nummer: 22381-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR