4,4'-Anisoin 95%
Lieferant: Thermo Scientific
Synonyme:
2-Hydroxy-1,2-bis(4-methoxyphenyl)ethanone, Anisoin
104841000
104840250
ACRO104841000EA
109
EUR
ACRO104841000
ACRO104840250
4,4'-Anisoin 95%
4,4'-Anisoin
Formel:
C₁₆H₁₆O₄ Molecular Weight: 272,30 g/mol Schmelzpunkt: 109…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008411 CAS-Nummer: 119-52-8 EINECS: 204-330-8 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 108 to 115 °C |
HPLC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...