5-Cumyl-o-anisidin Hydrochlorid
Lieferant: FLUOROCHEM
018466-1G
018466-5G
FLUO018466-1GEA
0
EUR
FLUO018466-1G
FLUO018466-5G
5-Cumyl-o-anisidin Hydrochlorid
5-Cumyl-o-anisidin Hydrochlorid
Formel:
C₁₆H₂₀ClNO |
MDL:
MFCD00270101 CAS-Nummer: 58999-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...