Salicylaldehydazin

Lieferant: FLUOROCHEM

Synonyme: Salicylaldazine

005449-10G 005449-25G
FLUO005449-10GEA 97.5 EUR
FLUO005449-10G FLUO005449-25G
Salicylaldehydazin
Salicylaldehydazin
Formel: C₁₄H₁₂N₂O₂
Molecular Weight: 240,26 g/mol
MDL: MFCD00043496
CAS-Nummer: 959-36-4
EINECS: 213-500-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR