3,5-Dimethyl-4-ethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Ethoxy-3,5-dimethylbenzeneboronic acid, 4-Ethoxy-3,5-dimethylphenylboronic acid, (4-Ethoxy-3,5-dimethylphenyl)boronic acid, 3,5-Dimethyl-4-ethoxybenzeneboronic acid

BB-2156-1G BB-2156-25G BB-2156-5G
COBBBB-2156-1GEA 37.5 EUR
COBBBB-2156-1G COBBBB-2156-25G COBBBB-2156-5G
3,5-Dimethyl-4-ethoxyphenylboronsäure
3,5-Dimethyl-4-ethoxyphenylboronsäure
Formel: C₁₀H₁₅BO₃
Molecular Weight: 194,04 g/mol
MDL: MFCD02179463
CAS-Nummer: 850568-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR