2,4-Bis(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2,4-Bis(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-3518-5G PN-3518-1G
COBBPN-3518-5GEA 286 EUR
COBBPN-3518-5G COBBPN-3518-1G
2,4-Bis(trifluormethyl)phenylboronsäurepinakolester
2,4-Bis(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₅BF₆O₂
Molecular Weight: 340.700 g/mol
MDL: MFCD09953467
CAS-Nummer: 1073353-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR