3-(Methoxycarbonyl)-5-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzoate

PN-5518-1G PN-5518-250M
COBBPN-5518-1GEA 800 EUR
COBBPN-5518-1G COBBPN-5518-250M
3-(Methoxycarbonyl)-5-(trifluormethyl)phenylboronsäurepinakolester
3-(Methoxycarbonyl)-5-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₅H₁₈BF₃O₄
Molecular Weight: 330.1072 g/mol
MDL: MFCD12026075
CAS-Nummer: 1150271-61-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR