Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3243-25G TB-3243-5G TB-3243-1G
COBBTB-3243-25GEA 0 EUR
COBBTB-3243-25G COBBTB-3243-5G COBBTB-3243-1G
Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat
Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat
Formel: C₈H₃BF₉K
MDL: MFCD03701625
CAS-Nummer: 166328-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR