Benzo[b]thiophen-2-carbaldehyd ≥97%
Lieferant: Thermo Scientific
Synonyme:
Benzo[b]thiophene-2-carbaldehyde
L19345.03
L19345.06
L19345.03EA
63.8
EUR
L19345.03
L19345.06
Benzo[b]thiophen-2-carbaldehyd ≥97%
Benzo[b]thiophen-2-carbaldehyd
Purity: 97%
CAS No.: 3541-37-5
Molecular Formula: C9H6OS
Formula Weight: 162.21
Melting Point: 30-32°
Boiling Point: 135-136°/8mm
MDL No.: MFCD01075041
Formel:
C₉H₆OS Molecular Weight: 162,21 g/mol Siedepunkt: 135…136 °C (8 mmHg) Schmelzpunkt: 30…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075041 CAS-Nummer: 3541-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...