6-Morpholinonicotinsäure
Lieferant: BIONET RESEARCH
2Y-0709.500
2Y-0709.5
2Y-0709.1
2Y-0709.10
BIOT2Y-0709.500EA
0
EUR
BIOT2Y-0709.500
BIOT2Y-0709.5
BIOT2Y-0709.1
BIOT2Y-0709.10
6-Morpholinonicotinsäure
6-Morpholinonicotinsäure
Formel:
C₁₀H₁₂N₂O₃ Molecular Weight: 208.213 g/mol |
MDL:
MFCD02682034 CAS-Nummer: 120800-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...