Kalium(2-(benzyloxy)phenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
TB-3209-1G
TB-3209-5G
COBBTB-3209-1GEA
149
EUR
COBBTB-3209-1G
COBBTB-3209-5G
Kalium(2-(benzyloxy)phenyl)trifluoroborat
Kalium(2-(benzyloxy)phenyl)trifluoroborat
Formel:
C₁₃H₁₁BF₃KO Molecular Weight: 290,13 g/mol |
MDL:
MFCD04115742 CAS-Nummer: 850623-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...