Kalium(3-(benzyloxy)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium (3-(benzyloxy)phenyl)trifluoroborate

TB-3221-1G TB-3221-25G TB-3221-5G
COBBTB-3221-1GEA 47.5 EUR
COBBTB-3221-1G COBBTB-3221-25G COBBTB-3221-5G
Kalium(3-(benzyloxy)phenyl)trifluoroborat
Kalium(3-(benzyloxy)phenyl)trifluoroborat
Formel: C₁₃H₁₁BF₃KO
Molecular Weight: 290.1300 g/mol
MDL: MFCD04115751
CAS-Nummer: 850623-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR