2,4-Dichlor-5-[(3-fluorbenzyl)oxy]anilin

Lieferant: BIONET RESEARCH

4T-0294.500 4T-0294.1 4T-0294.5
BIOT4T-0294.500EA 0 EUR
BIOT4T-0294.500 BIOT4T-0294.1 BIOT4T-0294.5
2,4-Dichlor-5-[(3-fluorbenzyl)oxy]anilin
2,4-Dichlor-5-[(3-fluorbenzyl)oxy]anilin
Formel: C₁₃H₁₀Cl₂FNO
MDL: MFCD03425777
CAS-Nummer: 439095-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR