4-Phenylbenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
4-Biphenylcarboxylic acid hydrazide
018959-10G
018959-25G
FLUO018959-10GEA
264
EUR
FLUO018959-10G
FLUO018959-25G
4-Phenylbenzhydrazid
4-Phenylbenzhydrazid
Formel:
C₁₃H₁₂N₂O Molecular Weight: 212.2472 g/mol |
MDL:
MFCD00017078 CAS-Nummer: 18622-23-6 EINECS: 242-451-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...