1-Benzyl-1-phenylhydrazin Hydrochlorid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Benzyl-N-phenylhydrazine hydrochloride

A17367.06 A17367.14
A17367.06EA 37.7 EUR
A17367.06 A17367.14
1-Benzyl-1-phenylhydrazin Hydrochlorid ≥98%
1-Benzyl-1-phenylhydrazin Hydrochlorid

Purity: 98+% CAS No.: 5705-15-7 Molecular Formula: C13H14N2.HCl Formula Weight: 234.73 Melting Point: 178-181° Beilstein Reference: 3707186

Formel: C₁₃H₁₅ClN₂
Molecular Weight: 234,73 g/mol
Schmelzpunkt: 178…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00050690
CAS-Nummer: 5705-15-7
EINECS: 227-195-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR