Calciumdisilicid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Calcium silicide
21240-1KG-F
21240-250G-F
SIAL21240-1KG-FEA
115
EUR
SIAL21240-1KG-F
SIAL21240-250G-F
Calciumdisilicid, Sigma-Aldrich®
Calcium disilicide
Formel:
CaSi₂ Molecular Weight: 96,25 g/mol |
MDL:
MFCD00015977 CAS-Nummer: 12013-56-8 EINECS: 234-588-7 UN: 1405 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...