Tris(trimethylsilyl)borat

Lieferant: CHEMPUR SILANE

Synonyme: Tris(trimethylsiloxy)borate

SILS21250.10 SILS21250.25
SILNSILS21250.10EA 0 EUR
SILNSILS21250.10 SILNSILS21250.25
Tris(trimethylsilyl)borat
Tris(trimethylsilyl)borat
Formel: C₉H₂₇BO₃Si₃
Molecular Weight: 278.376 g/mol
MDL: MFCD00051588
CAS-Nummer: 4325-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR