Hexaborsilicid

Lieferant: CHEMPUR SILANE

SILS14525.10 SILS14525.25
SILNSILS14525.10EA 0 EUR
SILNSILS14525.10 SILNSILS14525.25
Hexaborsilicid
Hexaboron silicide
Formel: B₆Si
MDL: MFCD00180907
CAS-Nummer: 12008-29-6
EINECS: 234-535-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR