Cerdisilicid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
Synonyme:
Cerium silicide
35836.06
35836.14
35836.06EA
95.3
EUR
35836.06
35836.14
Cerdisilicid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Cerium disilicide
Purity: 99.9%
CAS No.: 12014-85-6
Molecular Formula: CeSi2
Formula Weight: 196.29
Physical Form: 3-15mm Pieces
Density: 5.31
Appearance: Grey
Odour: Odourless
Melting Point: 1620°
MDL No.: MFCD00168082
Formel:
CeSi₂ Molecular Weight: 196,29 g/mol Schmelzpunkt: 1620 °C Dichte: 5,31 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168082 CAS-Nummer: 12014-85-6 EINECS: 234-602-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...