Titandisilicid ≥99,5% (Metall-Basis), Pulver
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Titansilicid
13088.18
13088.30
13088.18EA
169
EUR
13088.18
13088.30
Titandisilicid ≥99,5% (Metall-Basis), Pulver
Titanium disilicide
Purity: 99.5%
CAS No.: 12039-83-7
Molecular Formula: Si2Ti
Formula Weight: 104.07
Physical Form: -200 Mesh Powder
Appearance: Grey
Melting Point: 1760°
UN No.: 3178
MDL No.: MFCD00151541
Formel:
Si₂Ti Molecular Weight: 104.07 g/mol Schmelzpunkt: 1760 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151541 CAS-Nummer: 12039-83-7 EINECS: 234-904-3 UN: 3178 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...