Zirkoniumdisilicid ≥99,5% Metall-Basis ausgeschlossen Hf (max. 4% Hf)

Lieferant: Thermo Fisher Scientific

Synonyme: Zirconium silicide

36322.14 36322.22
36322.14EA 122 EUR
36322.14 36322.22
Zirkoniumdisilicid ≥99,5% Metall-Basis ausgeschlossen Hf (max. 4% Hf)
Zirconium disilicide

Purity: 99.5% CAS No.: 12039-90-6 Molecular Formula: ZrSi2 (may contain other phases) Formula Weight: 147.39 Physical Form: -325 Mesh Powder Density: 4.88 Appearance: Grey Odour: Odourless MDL No.: MFCD00151547

Formel: Si₂Zr
Molecular Weight: 147.39 g/mol
Dichte: 4.88
Lagertemperatur: Raumtemperatur
MDL: MFCD00151547
CAS-Nummer: 12039-90-6
EINECS: 234-911-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR