Niobdisilicid ≥99,85% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

13100.14 13100.22
13100.14EA 94.7 EUR
13100.14 13100.22
Niobdisilicid ≥99,85% (Metall-Basis)
Niobium disilicide

Purity: 99.85% CAS No.: 12034-80-9 Molecular Formula: NbSi2 Formula Weight: 149.08 Physical Form: -325 Mesh Powder Density: 5.7 Melting Point: 1950° MDL No.: MFCD00151537

Formel: NbSi₂
Molecular Weight: 149,08 g/mol
Schmelzpunkt: 1950 °C
Dichte: 5,7
Lagertemperatur: Raumtemperatur
MDL: MFCD00151537
CAS-Nummer: 12034-80-9
EINECS: 234-812-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR