Niobdisilicid

Lieferant: CHEMPUR SILANE

SILS12131.25 SILS12131.100
SILNSILS12131.25EA 0 EUR
SILNSILS12131.25 SILNSILS12131.100
Niobdisilicid
Niobium disilicide
Formel: NbSi₂
Molecular Weight: 149.08 g/mol
MDL: MFCD00151537
CAS-Nummer: 12034-80-9
EINECS: 234-812-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR