Tantaldisilicid
Lieferant: Thermo Fisher Scientific
Synonyme:
Tantalum silicide
44965.18
44965.09
44965.18EA
285
EUR
44965.18
44965.09
Tantaldisilicid
Tantalum disilicide
CAS No.: 12039-79-1
Molecular Formula: TaSi2
Formula Weight: 237.12
Physical Form: -325 Mesh Powder
Melting Point: 2200°
MDL No.: MFCD00049564
Formel:
Si₂Ta Molecular Weight: 237.118 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049564 CAS-Nummer: 12039-79-1 EINECS: 234-902-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...