Chromdisilicid ≥99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Chromium silicide
12151.18
12151.30
12151.18EA
66
EUR
12151.18
12151.30
Chromdisilicid ≥99% (Metall-Basis)
Chromium disilicide
Purity: 99+%
CAS No.: 12018-09-6
Molecular Formula: CrSi2
Formula Weight: 108.17
Physical Form: Powder
Sensitivity: Moisture Sensitive
MDL No.: MFCD00168070
Formel:
CrSi₂ Molecular Weight: 108,17 g/mol Schmelzpunkt: 1490 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168070 CAS-Nummer: 12018-09-6 EINECS: 234-633-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...