Molybdändisilicid (≥61% Mo) ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
Synonyme: Molybdenum silicide
11549.18 11549.30
11549.18EA 85.7 EUR
11549.18 11549.30
Molybdändisilicid (≥61% Mo) ≥99,5% (Metall-Basis)
Molybdenum disilicide
Purity: 99.5% CAS No.: 12136-78-6 Molecular Formula: MoSi2 Formula Weight: 152.13 Physical Form: -325 Mesh Powder Density: 6.31 Melting Point: 1870-2030° Solubility: Insoluble in acids and aqua regia, however, it is soluble in HF and HNO3 MDL No.: MFCD00151535
In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles
Formel: MoSi₂
Molecular Weight: 152,11 g/mol
Schmelzpunkt: 1870…2030 °C
Dichte: 6,31
Lagertemperatur: Raumtemperatur
MDL: MFCD00151535
CAS-Nummer: 12136-78-6
EINECS: 235-231-8