Molybdändisilicid

Lieferant: CHEMPUR SILANE

Synonyme: Molybdenum silicide

SILS12130.25 SILS12130.100
SILNSILS12130.25EA 0 EUR
SILNSILS12130.25 SILNSILS12130.100
Molybdändisilicid
Molybdenum disilicide
Formel: MoSi₂
Molecular Weight: 152.11 g/mol
MDL: MFCD00151535
CAS-Nummer: 12136-78-6
EINECS: 235-231-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR