Cobaltdisilicid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Cobalt silicide

13115.14 13115.22
13115.14EA 62.5 EUR
13115.14 13115.22
Cobaltdisilicid ≥99% (Metall-Basis)
Cobalt disilicide

Purity: 99% CAS No.: 12017-12-8 Molecular Formula: CoSi2 Formula Weight: 115.11 Physical Form: Powder Density: 5.30 Appearance: Grey Odour: Odourless Melting Point: 1277° MDL No.: MFCD00016021

Formel: CoSi₂
Molecular Weight: 115.11 g/mol
Schmelzpunkt: 1277 °C
Dichte: 5.30
Lagertemperatur: Raumtemperatur
MDL: MFCD00016021
CAS-Nummer: 12017-12-8
EINECS: 234-616-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR