Eisendisilicid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ferrosilicon, Iron silicide

14022.06 14022.14
14022.06EA 40.3 EUR
14022.06 14022.14
Eisendisilicid ≥99,9% (Metall-Basis)
Iron disilicide

Purity: 99.9% CAS No.: 12022-99-0 Molecular Formula: FeSi2 Formula Weight: 112.02 Physical Form: -20 Mesh Odour: Odourless MDL No.: MFCD00016087

Formel: FeSi₂
Molecular Weight: 112.02 g/mol
Schmelzpunkt: 1220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016087
CAS-Nummer: 12022-99-0
EINECS: 234-671-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR