4,7-Dibrom-3-hydroxy-2-naphthoesäure

Lieferant: FLUOROCHEM

Synonyme: 1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid

013143-5G 013143-25G
FLUO013143-5GEA 0 EUR
FLUO013143-5G FLUO013143-25G
4,7-Dibrom-3-hydroxy-2-naphthoesäure
4,7-Dibrom-3-hydroxy-2-naphthoesäure
Formel: C₁₁H₆Br₂O₃
Molecular Weight: 345,98 g/mol
MDL: MFCD00046367
CAS-Nummer: 1779-10-8
EINECS: 217-214-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR