4-Brom-1-fluor-2-naphthalinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Bromo-1-fluoronaphthalen-2-yl)boronic acid
BB-3583-5G
BB-3583-1G
COBBBB-3583-5GEA
1180
EUR
COBBBB-3583-5G
COBBBB-3583-1G
4-Brom-1-fluor-2-naphthalinboronsäure
4-Bromo-1-fluoro-2-naphthaleneboronic acid
Formel:
C₁₀H₇BBrFO₂ Molecular Weight: 268,8748 g/mol |
MDL:
MFCD09027236 CAS-Nummer: 913836-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...