2-(Bromacetyl)naphthalin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone

B23960.09 B23960.18 B23960.30
B23960.09EA 50 EUR
B23960.09 B23960.18 B23960.30
2-(Bromacetyl)naphthalin ≥98%
2-(Bromacetyl)naphthalin

Purity: 98% CAS No.: 613-54-7 Molecular Formula: C12H9BrO Formula Weight: 249.11 Melting Point: 80-85° Beilstein Reference: 639153 UN No.: 3261 MDL No.: MFCD00004109

Formel: C₁₂H₉BrO
Molecular Weight: 249.11 g/mol
Schmelzpunkt: 80…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004109
CAS-Nummer: 613-54-7
EINECS: 210-348-7
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR