2-(Bromacetyl)naphthalin

Lieferant: BIONET RESEARCH

Synonyme: α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone

7K-900.500 7K-900.1 7K-900.5 7K-900.10
BIOT7K-900.500EA 0 EUR
BIOT7K-900.500 BIOT7K-900.1 BIOT7K-900.5 BIOT7K-900.10
2-(Bromacetyl)naphthalin
2-(Bromacetyl)naphthalin
Formel: C₁₂H₉BrO
Molecular Weight: 249,11 g/mol
MDL: MFCD00004109
CAS-Nummer: 613-54-7
EINECS: 210-348-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR