1-Brom-4-(hydroxymethyl)naphthalin

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromonaphthalen-1-yl)methanol

HC-3404-1G HC-3404-250M
COBBHC-3404-1GEA 864 EUR
COBBHC-3404-1G COBBHC-3404-250M
1-Brom-4-(hydroxymethyl)naphthalin
1-Bromo-4-(hydroxymethyl)naphthalene
Formel: C₁₁H₉BrO
MDL: MFCD06658378
CAS-Nummer: 56052-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR