3,5-Dibrompyridin-N-oxid

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dibromopyridine1-oxide

PY-1352-5G PY-1352-25G PY-1352-1G
COBBPY-1352-5GEA 124 EUR
COBBPY-1352-5G COBBPY-1352-25G COBBPY-1352-1G
3,5-Dibrompyridin-N-oxid
3,5-Dibrompyridin-N-oxid
Formel: C₅H₃Br₂NO
Molecular Weight: 252,8914 g/mol
MDL: MFCD00160750
CAS-Nummer: 2402-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR