3-Brompyridin-N-oxid

Lieferant: CHEMPUR

69-0009.1 69-0009.5
CHMP69-0009.1EA 0 EUR
CHMP69-0009.1 CHMP69-0009.5
3-Brompyridin-N-oxid
3-Brompyridin-N-oxid
Formel: C₅H₄BrNO
MDL: MFCD01646056
CAS-Nummer: 2402-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR