3,5-Dichlorpyridin-N-oxid

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dichloropyridine1-oxide

PY-1354-5G PY-1354-1G
COBBPY-1354-5GEA 130 EUR
COBBPY-1354-5G COBBPY-1354-1G
3,5-Dichlorpyridin-N-oxid
3,5-Dichlorpyridin-N-oxid
Formel: C₅H₃Cl₂NO
Molecular Weight: 163,9894 g/mol
MDL: MFCD00955620
CAS-Nummer: 15177-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR