3,5-Difluorpyridin-N-oxid

Lieferant: COMBI-BLOCKS

PY-1359-5G PY-1359-1G
COBBPY-1359-5GEA 595 EUR
COBBPY-1359-5G COBBPY-1359-1G
3,5-Difluorpyridin-N-oxid
3,5-Difluorpyridin-N-oxid
Formel: C₅H₃F₂NO
MDL: MFCD16036156
CAS-Nummer: 210169-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR